Learn More
Biomatik Corporation Recombinant Human Zymogen granule protein 16 homolog B (ZG16B), 100UG
Catalog No. 501504725
Encompass_Preferred
Recombinant Human Zymogen granule protein 16 homolog B (ZG16B) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ZG16B. Target Synonyms: ZG16B; UNQ773/PRO1567Zymogen granule protein 16 homolog B. Accession Number: Q96DA0. Expression Region: 53~208aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 19.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4725 Supplier Biomatik Corporation Supplier No. RPC20326
Recombinant Human Zymogen granule protein 16 homolog B (ZG16B) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ZG16B. Target Synonyms: ZG16B; UNQ773/PRO1567Zymogen granule protein 16 homolog B. Accession Number: Q96DA0. Expression Region: 53~208aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 19.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.