Learn More
Biomatik Corporation Recombinant Influenza A virus Hemagglutinin (HA), partial, 100UG
Supplier:  Biomatik Corporation RPC20327
Encompass_Preferred
Recombinant Influenza A virus Hemagglutinin (HA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Influenza A virus (strain A/Hickox/1940 H1N1). Target Name: HA. Target Synonyms: Cleaved into the following 2 chains:Hemagglutinin HA1 chainHemagglutinin HA2 chain. Accession Number: Q0HD60. Expression Region: 18~343aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 52.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4726