Learn More
Biomatik Corporation Recombinant Human High mobility group protein B1 (Hmgb1), 100UG
Catalog No. 501504727
Encompass_Preferred
Recombinant Human High mobility group protein B1 (HMGB1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: Hmgb1. Target Synonyms: High mobility group protein 1HMG-1. Accession Number: P09429. Expression Region: 2~215aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4727 Supplier Biomatik Corporation Supplier No. RPC20328
Recombinant Human High mobility group protein B1 (HMGB1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: Hmgb1. Target Synonyms: High mobility group protein 1HMG-1. Accession Number: P09429. Expression Region: 2~215aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.