Learn More
Biomatik Corporation Recombinant Malus domestica Major allergen Mal d 1 (MALD1), 100UG
Supplier:  Biomatik Corporation RPC20332
Encompass_Preferred
Recombinant Malus domestica Major allergen Mal d 1 (MALD1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Malus domestica (Apple) (Pyrus malus). Target Name: MALD1. Target Synonyms: Allergen Mal d IAllergen: Mal d 1. Accession Number: P43211. Expression Region: 2~159aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 33.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4729