Learn More
Biomatik Corporation Recombinant Calloselasma rhodostoma Snaclec rhodocytin subunit alpha, 100UG
Catalog No. 501504731
Encompass_Preferred
Recombinant Calloselasma rhodostoma Snaclec rhodocytin subunit alpha is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Calloselasma rhodostoma (Malayan pit viper) (Agkistrodon rhodostoma). Target Name: Calloselasma rhodostoma Snaclec rhodocytin subunit alpha. Target Synonyms: Aggretin alpha chainRhodoaggretin subunit alpha. Accession Number: Q9I841. Expression Region: 1~136aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 31.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4731 Supplier Biomatik Corporation Supplier No. RPC20337
Recombinant Calloselasma rhodostoma Snaclec rhodocytin subunit alpha is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Calloselasma rhodostoma (Malayan pit viper) (Agkistrodon rhodostoma). Target Name: Calloselasma rhodostoma Snaclec rhodocytin subunit alpha. Target Synonyms: Aggretin alpha chainRhodoaggretin subunit alpha. Accession Number: Q9I841. Expression Region: 1~136aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 31.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.