Learn More
Biomatik Corporation Recombinant Human MORC family CW-type zinc finger protein 3 (MORC3), partial, 100UG
Catalog No. 501504733
Encompass_Preferred
Recombinant Human MORC family CW-type zinc finger protein 3 (MORC3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MORC3. Target Synonyms: Nuclear matrix protein 21 Zinc finger CW-type coiled-coil domain protein 3. Accession Number: Q14149. Expression Region: 1~290aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 48.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4733 Supplier Biomatik Corporation Supplier No. RPC20339
Recombinant Human MORC family CW-type zinc finger protein 3 (MORC3), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MORC3. Target Synonyms: Nuclear matrix protein 21 Zinc finger CW-type coiled-coil domain protein 3. Accession Number: Q14149. Expression Region: 1~290aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 48.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.