Learn More
Biomatik Corporation Recombinant Human Semenogelin-1 (SEMG1), 100UG
Supplier:  Biomatik Corporation RPC20353
Encompass_Preferred
Recombinant Human Semenogelin-1 (SEMG1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: SEMG1. Target Synonyms: Cancer/testis antigen 103Semenogelin ISGICleaved into the following 3 chains:Alpha-inhibin-92Alpha-inhibin-31Seminal basic protein. Accession Number: P04279. Expression Region: 24~402aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 58.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4741