Learn More
Biomatik Corporation Recombinant Human parvovirus B19 Non-capsid protein NS-1 (NS1), partial, 100UG
Supplier:  Biomatik Corporation RPC20356
Encompass_Preferred
Recombinant Human parvovirus B19 Non-capsid protein NS-1 (NS1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human parvovirus B19 (isolate AU) (HPV B19). Target Name: NS1. Target Synonyms: NCVP1Non-structural protein NS1. Accession Number: P07298. Expression Region: 1~255aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 44.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4744