Learn More
Biomatik Corporation Recombinant Antifungal protein (afp), 100UG
Catalog No. 501504747
Encompass_Preferred
Recombinant Aspergillus giganteus Antifungal protein (afp) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Aspergillus giganteus. Target Name: afp. Target Synonyms: afpAntifungal protein. Accession Number: P17737. Expression Region: 44~94aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 19.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4747 Supplier Biomatik Corporation Supplier No. RPC20359
Recombinant Aspergillus giganteus Antifungal protein (afp) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Aspergillus giganteus. Target Name: afp. Target Synonyms: afpAntifungal protein. Accession Number: P17737. Expression Region: 44~94aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 19.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.