Learn More
Biomatik Corporation Recombinant Bacillus subtilis Penicillin-binding protein 3 (pbpC), partial, 100UG
Supplier:  Biomatik Corporation RPC20360
Encompass_Preferred
Recombinant Bacillus subtilis Penicillin-binding protein 3 (pbpC), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bacillus subtilis (strain 168). Target Name: pbpC. Target Synonyms: PBP 3PSPB20Penicillin-binding protein C. Accession Number: P42971. Expression Region: 21~240aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 29.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4748