Learn More
Biomatik Corporation Recombinant Rat Monoglyceride lipase (Mgll), 100UG
Supplier:  Biomatik Corporation RPC20364
Encompass_Preferred
Recombinant Rat Monoglyceride lipase (Mgll) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Mgll. Target Synonyms: MGLMonoacylglycerol lipaseMAGL. Accession Number: Q8R431. Expression Region: 1~303aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 34.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4750