Learn More
Biomatik Corporation Recombinant Raphanus sativus Defensin-like protein 2 (AFP2), 100UG
Supplier:  Biomatik Corporation RPC20365
Encompass_Preferred
Recombinant Raphanus sativus Defensin-like protein 2 (AFP2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Raphanus sativus (Radish). Target Name: AFP2. Target Synonyms: Cysteine-rich antifungal protein 2AFP2RAFP2. Accession Number: P30230. Expression Region: 30~80aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 21.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4751