Learn More
Biomatik Corporation Recombinant Human Putative 40S ribosomal protein S10-like (RPS10P5), 100UG
Supplier:  Biomatik Corporation RPC20371
Encompass_Preferred
Recombinant Human Putative 40S ribosomal protein S10-like (RPS10P5) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: RPS10P5. Target Synonyms: RPS10P5; RPS10LPutative 40S ribosomal protein S10-like. Accession Number: Q9NQ39. Expression Region: 1~176aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 36.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4754