Learn More
Biomatik Corporation Recombinant Yersinia enterocolitica Invasin, partial, 100UG
Catalog No. 501504756
Encompass_Preferred
Recombinant Yersinia enterocolitica Invasin is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Yersinia enterocolitica. Target Name: Yersinia enterocolitica Invasin. Target Synonyms: Invasin. Accession Number: P19196. Expression Region: 651~835aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 36.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4756 Supplier Biomatik Corporation Supplier No. RPC20373
Recombinant Yersinia enterocolitica Invasin is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Yersinia enterocolitica. Target Name: Yersinia enterocolitica Invasin. Target Synonyms: Invasin. Accession Number: P19196. Expression Region: 651~835aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 36.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.