Learn More
Biomatik Corporation Recombinant Acinetobacter baylyi Catechol 1,2-dioxygenase (catA), 100UG
Catalog No. 501504758
Encompass_Preferred
Recombinant Acinetobacter baylyi Catechol 1, 2-dioxygenase (catA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Acinetobacter baylyi. Target Name: catA. Target Synonyms: 1,2-CTD. Accession Number: P07773. Expression Region: 1~311aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 50.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4758 Supplier Biomatik Corporation Supplier No. RPC20376
Recombinant Acinetobacter baylyi Catechol 1, 2-dioxygenase (catA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Acinetobacter baylyi. Target Name: catA. Target Synonyms: 1,2-CTD. Accession Number: P07773. Expression Region: 1~311aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 50.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.