Learn More
Biomatik Corporation Recombinant Mouse Fibroblast growth factor 21 (Fgf21), 100UG
Supplier:  Biomatik Corporation RPC20039
Encompass_Preferred
Recombinant Mouse Fibroblast growth factor 21 (Fgf21) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Fgf21. Target Synonyms: Fgf21Fibroblast growth factor 21; FGF-21. Accession Number: Q9JJN1. Expression Region: 29~210aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 23.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4553