Learn More
Biomatik Corporation Recombinant Glycine max Ferritin-2, chloroplastic, 100UG
Supplier:  Biomatik Corporation RPC20389
Encompass_Preferred
Recombinant Glycine max Ferritin-2, chloroplastic is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Glycine max (Soybean) (Glycine hispida). Target Name: Glycine max Ferritin-2, chloroplastic. Target Synonyms: SFerH-2. Accession Number: Q94IC4. Expression Region: 52~257aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 39.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4770