Learn More
Biomatik Corporation Recombinant Human Dystonin (DST), partial, 100UG
Supplier:  Biomatik Corporation RPC20393
Encompass_Preferred
Recombinant Human Dystonin (DST), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DST. Target Synonyms: 230 kDa bullous pemphigoid antigen230/240 kDa bullous pemphigoid antigenBullous pemphigoid antigen 1BPABullous pemphigoid antigen. Accession Number: Q03001. Expression Region: 1~195aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 39.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4772