Learn More
Biomatik Corporation Recombinant Chlamydia trachomatis Small cysteine-rich outer membrane protein OmcA (OmcA), 100UG
Supplier:  Biomatik Corporation RPC20395
Encompass_Preferred
Recombinant Chlamydia trachomatis Small cysteine-rich outer membrane protein OmcA (OmcA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Chlamydia trachomatis (strain D/UW-3/Cx). Target Name: omcA. Target Synonyms: 9 kDa cysteine-rich lipoprotein9 kDa-CRP. Accession Number: P0CC05. Expression Region: 19~88aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 23.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4774