Learn More
Biomatik Corporation Recombinant Arabidopsis thaliana 2-Cys peroxiredoxin BAS1, chloroplastic (BAS1), 100UG
Catalog No. 501504776
Encompass_Preferred
Recombinant Arabidopsis thaliana 2-Cys peroxiredoxin BAS1, chloroplastic (BAS1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Arabidopsis thaliana (Mouse-ear cress). Target Name: BAS1. Target Synonyms: Thiol-specific antioxidant protein A. Accession Number: Q96291. Expression Region: 66~266aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 24.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4776 Supplier Biomatik Corporation Supplier No. RPC20398
Recombinant Arabidopsis thaliana 2-Cys peroxiredoxin BAS1, chloroplastic (BAS1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Arabidopsis thaliana (Mouse-ear cress). Target Name: BAS1. Target Synonyms: Thiol-specific antioxidant protein A. Accession Number: Q96291. Expression Region: 66~266aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 24.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.