Learn More
Biomatik Corporation Recombinant Mouse Nucleophosmin (Npm1), 100UG
Supplier:  Biomatik Corporation RPC20406
Encompass_Preferred
Recombinant Mouse Nucleophosmin (Npm1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Npm1. Target Synonyms: Nucleolar phosphoprotein B23Nucleolar protein NO38Numatrin. Accession Number: Q61937. Expression Region: 1~292aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 48.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4782