Learn More
Biomatik Corporation Recombinant Mouse Acetyl-coenzyme A synthetase, Cytoplasmic domain (Acss2), 100UG
Supplier:  Biomatik Corporation RPC20408
Encompass_Preferred
Recombinant Mouse Acetyl-coenzyme A synthetase, Cytoplasmic domain (Acss2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Acss2. Target Synonyms: Acetate--CoA ligaseAcetyl-CoA synthetaseACSAceCS. Accession Number: Q9QXG4. Expression Region: 1~701aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 82.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4784