Learn More
Biomatik Corporation Recombinant Mouse Lithostathine-1 (Reg1), 100UG
Supplier:  Biomatik Corporation RPC20409
Encompass_Preferred
Recombinant Mouse Lithostathine-1 (Reg1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Reg1. Target Synonyms: Islet of Langerhans regenerating protein 1REG 1Pancreatic stone protein 1PSPPancreatic thread protein 1PTPRegenerating protein 1. Accession Number: P43137. Expression Region: 22~165aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 32.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4785