Learn More
Biomatik Corporation Recombinant Chicken Homeobox protein engrailed-1 (EN1), 100UG
Supplier:  Biomatik Corporation RPC20415
Encompass_Preferred
Recombinant Chicken Homeobox protein engrailed-1 (EN1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Chicken (Gallus). Target Name: EN1. Target Synonyms: Gg-En-1Homeobox protein en-1. Accession Number: Q05916. Expression Region: 1~333aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 48.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4788