Learn More
Biomatik Corporation Recombinant Pig Epididymal secretory glutathione peroxidase (GPX5), 100UG
Supplier:  Biomatik Corporation RPC20416
Encompass_Preferred
Recombinant Pig Epididymal secretory glutathione peroxidase (GPX5) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Pig (Sus scrofa; Porcine). Target Name: GPX5. Target Synonyms: Epididymis-specific glutathione peroxidase-like proteinEGLP. Accession Number: O18994. Expression Region: 22~219aa. Tag Info: N-Terminal 6Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 26.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4789