Learn More
Biomatik Corporation Recombinant Human 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGCR), partial, 100UG
Catalog No. 501504556
Encompass_Preferred
Recombinant Human 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGCR), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HMGCR. Target Synonyms: 3 hydroxy 3 methylglutaryl CoA reductase; 3 hydroxy 3 methylglutaryl Coenzyme A reductase; 3 hydroxymethylglutaryl CoA reductase; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH). Accession Number: P04035. Expression Region: 588~887aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 36kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4556 Supplier Biomatik Corporation Supplier No. RPC20050
Recombinant Human 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGCR), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HMGCR. Target Synonyms: 3 hydroxy 3 methylglutaryl CoA reductase; 3 hydroxy 3 methylglutaryl Coenzyme A reductase; 3 hydroxymethylglutaryl CoA reductase; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH). Accession Number: P04035. Expression Region: 588~887aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 36kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.