Learn More
Biomatik Corporation Recombinant Lolium perenne Major pollen allergen Lol p 5a (LOLPIB), 100UG
Catalog No. 501504791
Encompass_Preferred
Recombinant Lolium perenne Major pollen allergen Lol p 5a (LOLPIB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Lolium perenne. Target Name: LOLPIB. Target Synonyms: Allergen Lol p IbAllergen Lol p VaAllergen: Lol p 5a. Accession Number: Q40240. Expression Region: 26~307aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 48.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4791 Supplier Biomatik Corporation Supplier No. RPC20418
Recombinant Lolium perenne Major pollen allergen Lol p 5a (LOLPIB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Lolium perenne. Target Name: LOLPIB. Target Synonyms: Allergen Lol p IbAllergen Lol p VaAllergen: Lol p 5a. Accession Number: Q40240. Expression Region: 26~307aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 48.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.