Learn More
Biomatik Corporation Recombinant Horse Major allergen Equ c 1, 100UG
Supplier:  Biomatik Corporation RPC20421
Encompass_Preferred
Recombinant Horse Major allergen Equ c 1 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Horse (Equus caballus; Equine). Target Name: Horse Major allergen Equ c 1. Target Synonyms: Allergen: Equ c 1. Accession Number: Q95182. Expression Region: 16~187aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 22.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4793