Learn More
Biomatik Corporation Recombinant Arabidopsis thaliana Osmotin-like protein OSM34 (OSM34), 100UG
Supplier:  Biomatik Corporation RPC20422
Encompass_Preferred
Recombinant Arabidopsis thaliana Osmotin-like protein OSM34 (OSM34) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Arabidopsis thaliana (Mouse-ear cress). Target Name: OSM34. Target Synonyms: OSL3_ARATH; OSM34; Osmotin-like protein OSM34. Accession Number: P50700. Expression Region: 23~244aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 44.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4794