Learn More
Biomatik Corporation Recombinant Streptococcus pyogenes Streptopain (speB), 100UG
Supplier:  Biomatik Corporation RPC20423
Encompass_Preferred
Recombinant Streptococcus pyogenes Streptopain (speB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Streptococcus pyogenes. Target Name: speB. Target Synonyms: Exotoxin type BSPE BStreptococcal cysteine proteinaseStreptococcus peptidase ASPP. Accession Number: P0C0J0. Expression Region: 146~398aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 43.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4795