Learn More
Biomatik Corporation Recombinant Triangle waterfern Cyanovirin-N homolog, 100UG
Supplier:  Biomatik Corporation RPC20424
Encompass_Preferred
Recombinant Ceratopteris richardii Cyanovirin-N homolog is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Ceratopteris richardii (Triangle waterfern). Target Name: Ceratopteris richardii Cyanovirin-N homolog. Target Synonyms: Cyanovirin-N homolog; CV-N homolog. Accession Number: P86326. Expression Region: 28~142aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 14.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4796