Learn More
Biomatik Corporation Recombinant Shigella flexneri Invasin IpaD (IpaD), 100UG
Catalog No. 501504799
Encompass_Preferred
Recombinant Shigella flexneri Invasin IpaD (IpaD) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Shigella flexneri. Target Name: ipaD. Target Synonyms: 36 kDa membrane antigen. Accession Number: P18013. Expression Region: 1~332aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 56.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4799 Supplier Biomatik Corporation Supplier No. RPC20428
Recombinant Shigella flexneri Invasin IpaD (IpaD) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Shigella flexneri. Target Name: ipaD. Target Synonyms: 36 kDa membrane antigen. Accession Number: P18013. Expression Region: 1~332aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 56.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.