Learn More
Biomatik Corporation Recombinant Escherichia coli O157:H7 Outer membrane protein C (ompC), 100UG
Supplier:  Biomatik Corporation RPC20429
Encompass_Preferred
Recombinant Escherichia coli O157:H7 Outer membrane protein C (ompC) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli O157:H7. Target Name: ompC. Target Synonyms: Outer membrane protein 1BPorin OmpC. Accession Number: Q8XE41. Expression Region: 22~367aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 58.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4800