Learn More
Biomatik Corporation Recombinant Clostridium perfringens Perfringolysin O (pfo), 100UG
Catalog No. 501504802
Encompass_Preferred
Recombinant Clostridium perfringens Perfringolysin O (pfo) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Clostridium perfringens (strain 13 / Type A). Target Name: pfo. Target Synonyms: Theta-toxinThiol-activated cytolysin. Accession Number: P0C2E9. Expression Region: 29~500aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 68.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4802 Supplier Biomatik Corporation Supplier No. RPC20435
Recombinant Clostridium perfringens Perfringolysin O (pfo) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Clostridium perfringens (strain 13 / Type A). Target Name: pfo. Target Synonyms: Theta-toxinThiol-activated cytolysin. Accession Number: P0C2E9. Expression Region: 29~500aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 68.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.