Learn More
Biomatik Corporation Recombinant Cynodon dactylon Major pollen allergen Cyn d 1 (CYND1), 100UG
Catalog No. 501504807
Encompass_Preferred
Recombinant Cynodon dactylon Major pollen allergen Cyn d 1 (CYND1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Cynodon dactylon (Bermuda grass) (Panicum dactylon). Target Name: CYND1. Target Synonyms: CYND1; Major pollen allergen Cyn d 1; allergen Cyn d 1. Accession Number: O04701. Expression Region: 1~246aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 31.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4807 Supplier Biomatik Corporation Supplier No. RPC20442
Recombinant Cynodon dactylon Major pollen allergen Cyn d 1 (CYND1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Cynodon dactylon (Bermuda grass) (Panicum dactylon). Target Name: CYND1. Target Synonyms: CYND1; Major pollen allergen Cyn d 1; allergen Cyn d 1. Accession Number: O04701. Expression Region: 1~246aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 31.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.