Learn More
Biomatik Corporation Recombinant Renilla reniformis Coelenterazine h 2-monooxygenase, 100UG
Supplier:  Biomatik Corporation RPC20444
Encompass_Preferred
Recombinant Renilla reniformis Coelenterazine h 2-monooxygenase is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Renilla reniformis (Sea pansy). Target Name: Renilla reniformis Coelenterazine h 2-monooxygenase. Target Synonyms: Renilla-type luciferase. Accession Number: P27652. Expression Region: 1~311aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 56kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4808