Learn More
Biomatik Corporation Recombinant Human Chitinase-3-like protein 1 (CHI3L1), 100UG
Supplier:  Biomatik Corporation RPC20445
Encompass_Preferred
Recombinant Human Chitinase-3-like protein 1 (CHI3L1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CHI3L1. Target Synonyms: 39 kDa synovial protein Cartilage glycoprotein 39. Accession Number: P36222. Expression Region: 22~383aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 60.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4809