Learn More
Biomatik Corporation Recombinant Human Interferon alpha-14 (IFNA14), 100UG
Catalog No. 501504558
Encompass_Preferred
Recombinant Human Interferon alpha-14 (IFNA14) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IFNA14. Target Synonyms: Interferon alpha-H. Accession Number: P01570. Expression Region: 24~189aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 35.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4558 Supplier Biomatik Corporation Supplier No. RPC20052
Recombinant Human Interferon alpha-14 (IFNA14) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IFNA14. Target Synonyms: Interferon alpha-H. Accession Number: P01570. Expression Region: 24~189aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 35.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.