Learn More
Biomatik Corporation Recombinant Pseudomonas aeruginosa UDP-3-O-acyl-N-acetylglucosamine deacetylase (lpxC), 100UG
Supplier:  Biomatik Corporation RPC20446
Encompass_Preferred
Recombinant Pseudomonas aeruginosa UDP-3-O-acyl-N-acetylglucosamine deacetylase (lpxC) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Pseudomonas aeruginosa (strain ATCC 15692 / DSM 22644 / CIP 104116 / JCM 14847 / LMG 12228 / 1C / PRS 101 / PAO1). Target Name: lpxC. Target Synonyms: UDP-3-O-[R-3-hydroxymyristoyl]-N-acetylglucosamine deacetylase. Accession Number: P47205. Expression Region: 1~303aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 49.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4810