Learn More
Biomatik Corporation Recombinant Salmonella typhimurium D-galactose-binding periplasmic protein (mglB), 100UG
Catalog No. 501504811
Encompass_Preferred
Recombinant Salmonella typhimurium D-galactose-binding periplasmic protein (mglB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Salmonella typhimurium (strain LT2 / SGSC1412 / ATCC 700720). Target Name: mglB. Target Synonyms: D-galactose/ D-glucose-binding protein. Accession Number: P23905. Expression Region: 24~332aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 35.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4811 Supplier Biomatik Corporation Supplier No. RPC20449
Recombinant Salmonella typhimurium D-galactose-binding periplasmic protein (mglB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Salmonella typhimurium (strain LT2 / SGSC1412 / ATCC 700720). Target Name: mglB. Target Synonyms: D-galactose/ D-glucose-binding protein. Accession Number: P23905. Expression Region: 24~332aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 35.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.