Learn More
Biomatik Corporation Recombinant Human Histone deacetylase 6 (HDAC6), partial, 100UG
Supplier:  Biomatik Corporation RPC20451
Encompass_Preferred
Recombinant Human Histone deacetylase 6 (HDAC6), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HDAC6. Target Synonyms: CPBHM; FLJ16239; HD 6; HD6; HDAC 6; HDAC6; HDAC6_HUMAN; Histone deacetylase 6 (HD6); Histone deacetylase 6; JM 21; JM21; KIAA0901; OTTHUMP00000032398; OTTHUMP00000197663; PPP1R90; Protein phosphatase 1 regulatory subunit 90. Accession Number: Q9UBN7. Expression Region: 1~488aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 70.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4812