Learn More
Biomatik Corporation Recombinant Sheep Interleukin-4 (IL4), 100UG
Catalog No. 501504814
Encompass_Preferred
Recombinant Sheep Interleukin-4 (IL4) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Sheep (Ovis aries). Target Name: IL4. Target Synonyms: B-cell stimulatory factor 1. Accession Number: P30368. Expression Region: 25~135aa. Tag Info: N-Terminal 10Xhis-B2M-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 29.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4814 Supplier Biomatik Corporation Supplier No. RPC20454
Recombinant Sheep Interleukin-4 (IL4) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Sheep (Ovis aries). Target Name: IL4. Target Synonyms: B-cell stimulatory factor 1. Accession Number: P30368. Expression Region: 25~135aa. Tag Info: N-Terminal 10Xhis-B2M-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 29.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.