Learn More
Biomatik Corporation Recombinant Neisseria meningitidis serogroup B / serotype 15 Major outer membrane protein P.IB (porB), 100UG
Supplier:  Biomatik Corporation RPC20457
Encompass_Preferred
Recombinant Neisseria meningitidis serogroup B / serotype 15 Major outer membrane protein P.IB (porB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Neisseria meningitidis serogroup B / serotype 15 (strain H44/76). Target Name: porB. Target Synonyms: Class 3 proteinPorin. Accession Number: E6MZM0. Expression Region: 20~331aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 49.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4817