Learn More
Biomatik Corporation Recombinant Human Tripartite motif-containing protein 5 (TRIM5), 100UG
Supplier:  Biomatik Corporation RPC20458
Encompass_Preferred
Recombinant Human Tripartite motif-containing protein 5 (TRIM5) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TRIM5. Target Synonyms: RING finger protein 88. Accession Number: Q9C035. Expression Region: 1~493aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 70.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4818