Learn More
Biomatik Corporation Recombinant Human Galectin-9 (LGALS9), 100UG
Supplier:  Biomatik Corporation RPC20057
Encompass_Preferred
Recombinant Human Galectin-9 (LGALS9) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LGALS9. Target Synonyms: Ecalectin; Tumor antigen HOM-HD-21. Accession Number: O00182. Expression Region: 1~323aa. Tag Info: N-Terminal 6Xhis-Gst-Tagged. Theoretical MW: 65.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4559