Learn More
Biomatik Corporation Recombinant Human HLA class II histocompatibility antigen, DRB1-1 beta chain (HLA-DRB1), partial, 100UG
Catalog No. 501504822
Encompass_Preferred
Recombinant Human HLA class II histocompatibility antigen, DRB1 beta chain (HLA-DRB1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HLA-DRB1. Target Synonyms: MHC class II antigen DRB1*1. Accession Number: P04229. Expression Region: 30~227aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 42.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4822 Supplier Biomatik Corporation Supplier No. RPC20468
Recombinant Human HLA class II histocompatibility antigen, DRB1 beta chain (HLA-DRB1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HLA-DRB1. Target Synonyms: MHC class II antigen DRB1*1. Accession Number: P04229. Expression Region: 30~227aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 42.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.