Learn More
Biomatik Corporation Recombinant Brucella abortus Malate dehydrogenase (mdh), 100UG
Supplier:  Biomatik Corporation RPC20473
Encompass_Preferred
Recombinant Brucella abortus Malate dehydrogenase (mdh) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Brucella abortus (strain S19). Target Name: mdh. Target Synonyms: mdh; BAbS19_I18080; Malate dehydrogenase; EC 1.1.1.37. Accession Number: B2S881. Expression Region: 1~320aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 53.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4826