Learn More
Biomatik Corporation Recombinant Human Dynactin subunit 1 (DCTN1), partial, 100UG
Supplier:  Biomatik Corporation RPC20475
Encompass_Preferred
Recombinant Human Dynactin subunit 1 (DCTN1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DCTN1. Target Synonyms: 150 kDa dynein-associated polypeptideDAP-150. Accession Number: Q14203. Expression Region: 213~547aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 55.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4828