Learn More
Biomatik Corporation Recombinant Human Carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1), partial, 100UG
Supplier:  Biomatik Corporation RPC20480
Encompass_Preferred
Recombinant Human Carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CEACAM1. Target Synonyms: Biliary glycoprotein 1. Accession Number: P13688. Expression Region: 35~428aa. Tag Info: Tag-Free. Theoretical MW: 43.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4832