Learn More
Biomatik Corporation Recombinant Human Lactadherin (MFGE8), 100UG
Catalog No. 501504561
Encompass_Preferred
Recombinant Human Lactadherin (MFGE8) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MFGE8. Target Synonyms: Breast epithelial antigen BA46HMFGMFGMMilk fat globule-EGF factor 8; MFG-E8SED1. Accession Number: Q08431. Expression Region: 24~387aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 56.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4561 Supplier Biomatik Corporation Supplier No. RPC20059
Recombinant Human Lactadherin (MFGE8) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MFGE8. Target Synonyms: Breast epithelial antigen BA46HMFGMFGMMilk fat globule-EGF factor 8; MFG-E8SED1. Accession Number: Q08431. Expression Region: 24~387aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 56.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.